

4,6-Dichloro-5-methoxypyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5018-38-2
:
Formula:C₅H₄Cl₂N₂O
MDL Number:MFCD00087680
:179.01 g/mol
Melting Pt:54…58 °C
EINECS:255-699-1
:GHS05
:GHS07
Storage Temperature:Ambient