
Ethyl-3,4-dimethyl-1H-pyrrole-2-carboxylate
Catalog # APOSOR15759-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:167.21 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:938-75-0
Formula:C₉H₁₃NO₂
MDL Number:MFCD00030365