Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Octahydro-2H-pyrido[1,2-a]pyrazine
Octahydro-2H-pyrido[1,2-a]pyrazine 95%
Octahydro-2H-pyrido[1,2-a]pyrazine 95%
APOSOR15661-1G
:
Apollo Scientific
:
Octahydro-2H-pyrido[1,2-a]pyrazine 95%
APOSOR15661-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
140.23 g/mol
:
86…87 °C/11mmHg
:
Ambient
:
3267
:
8,II
:
GHS05
:
4430-75-5
:
C₈H₁₆N₂
:
MFCD01098257