
1-Bromo-2,4-dichlorobenzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1193-72-2
:
Formula:C₆H₃BrCl₂
MDL Number:MFCD00018333
:225.9 g/mol
Boiling Pt:235 °C
Melting Pt:26…30 °C
Density:1.89 g/cm³ (25 °C)
Flash Pt:98 °C
EINECS:214-778-6
:GHS07
Storage Temperature:Ambient