N-(10-Bromodecyl)phthalimide
Catalog # APOSOR1561-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:366.3 g/mol
Melting Pt:58…60 °C
Storage Temperature:Ambient
UN:3335
ADR:9,III
:GHS07
CAS Number:24566-80-1
Formula:C₁₈H₂₄BrNO₂
MDL Number:MFCD00060521