
1,3-Dimethoxybenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:151-10-0
:
Formula:C₈H₁₀O₂
MDL Number:MFCD00008384
:138.17 g/mol
Boiling Pt:217 °C
Melting Pt:–52 °C
Density:1.05 g/cm³ (25 °C)
Flash Pt:87 °C
EINECS:205-783-4
:GHS07
Storage Temperature:Ambient