Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,7-Dibromoheptane 95%

Toxic
New Product
Supplier:   Apollo Scientific
1,7-Dibromoheptane 95%
Supplier:  Apollo Scientific
New Product
Certificates
1,7-Dibromoheptane 95%

5 Options Available Below

Product Details & Documents
Synonyms:  Heptamethylene dibromide
  • CAS Number:4549-31-9
  • Formula:BrH₂C(CH₂)₅CH₂Br
  • MDL Number:MFCD00000274
  • Molecular Weight:258 g/mol
  • Boiling Pt:255 °C
  • Density:1.51 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:224-910-4
  • UN:2810
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 72,40 €
    72,40 €
    1 * 25 g
       72,40 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 231,00 €
    231,00 €
    1 * 100 g
       231,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 150,00 €
    1 150,00 €
    1 * 500 g
       1 150,00 €

    Specifications:
    New Product
    New Product