
Ethyl-2,5-dibromothiazole-4-carboxylate
Catalog # APOSOR15536-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:314.9800 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:208264-60-2
Formula:C₆H₅Br₂NO₂S
MDL Number:MFCD09265487