
Methyl-4-morpholinecarbodithioate
Catalog # APOSOR15505-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:177.29 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:62604-08-4
Formula:C₆H₁₁NOS₂
MDL Number:MFCD00277256