Order Entry
Export
ContactUsLinkComponent
1-(2-Furyl)-2,2-dimethyl-1,3-propanediol
Irritant
1-(2-Furyl)-2,2-dimethyl-1,3-propanediol
Catalog # APOSOR15495-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(2-Furyl)-2,2-dimethyl-1,3-propanediol
Catalog # APOSOR15495-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :170.21 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:4946-64-9
Formula:C₉H₁₄O₃
MDL Number:MFCD00720174