
6-Iodoimidazo[1,2-a]pyridine
Catalog # APOSOR15445-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:244.0300 g/mol
Melting Pt:130…131 °C
:GHS07
Storage Temperature:Ambient
CAS Number:426825-75-4
Formula:C₇H₅IN₂
MDL Number:MFCD06200985