0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
4-[(3,4-Dichlorobenzyl)oxy]benzenecarboxylic acid
4-[(3,4-Dichlorobenzyl)oxy]benzenecarboxylic acid
4-[(3,4-Dichlorobenzyl)oxy]benzenecarboxylic acid
APOSOR15439-1G
:
Apollo Scientific
:
4-[(3,4-Dichlorobenzyl)oxy]benzenecarboxylic acid
APOSOR15439-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
297.14 g/mol
:
210…214 °C
:
GHS07
:
Ambient
:
56442-18-3
:
C₁₄H₁₀Cl₂O₃
:
MFCD06200988