
4,5-Dibromo-1H-pyrrole-2-carboxylic acid
Catalog # APOSOR15435-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:268.9 g/mol
Melting Pt:>300 °C
:GHS07
Storage Temperature:Ambient
CAS Number:34649-21-3
Formula:C₅H₃Br₂NO₂
MDL Number:MFCD01348229
Melting Pt:>300 °C