Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Propyl-1,3-benzoxazol-5-amine
2-Propyl-1,3-benzoxazol-5-amine
2-Propyl-1,3-benzoxazol-5-amine
2-Propyl-1,3-benzoxazol-5-amine
Supplier:
Apollo Scientific
Certificates
Expand image
2-Propyl-1,3-benzoxazol-5-amine
Supplier:
Apollo Scientific
About this item
CAS Number:
886361-62-2
:
Formula:
C₁₀H₁₂N₂O
MDL Number:
MFCD06200946
:
176.22 g/mol
Storage Temperature:
Ambient