
2-Phenyl-1,3-thiazole-4-carbohydrazide
Catalog # APOSOR15428-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.27 g/mol
Melting Pt:136…138 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:7113-12-4
Formula:C₁₀H₉N₃OS
MDL Number:MFCD00174011