Order Entry
Export
ContactUsLinkComponent
Ethyl-2-(4-methoxyphenyl)-1,3-thiazole-4-carboxylate
Irritant
Ethyl-2-(4-methoxyphenyl)-1,3-thiazole-4-carboxylate
Catalog # APOSOR15332-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl-2-(4-methoxyphenyl)-1,3-thiazole-4-carboxylate
Catalog # APOSOR15332-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :263.3100 g/mol
Melting Pt:94…96 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:57677-79-9
Formula:C₁₃H₁₃NO₃S
MDL Number:MFCD06738343