Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Butyl-1,3-benzoxazol-4-amine
2-Butyl-1,3-benzoxazol-4-amine
2-Butyl-1,3-benzoxazol-4-amine
Irritant
2-Butyl-1,3-benzoxazol-4-amine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Butyl-1,3-benzoxazol-4-amine
Supplier:
Apollo Scientific
About this item
CAS Number:
886361-02-0
:
Formula:
C₁₁H₁₄N₂O
:
GHS07
Storage Temperature:
Ambient