
Methyl 1,6-dihydro-5-iodo-6-oxopyridine-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:885950-46-9
:
Formula:C₇H₆INO₃
MDL Number:MFCD08276965
:279.04 g/mol
Melting Pt:209…211 °C
:GHS07
Storage Temperature:Ambient