
2-Phenyl-1,3-thiazol-4-ol
Catalog # APOSOR15120-50MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:177.23 g/mol
Melting Pt:119…121 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:827-45-2
Formula:C₉H₇NOS
MDL Number:MFCD08444022