Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 6-methoxynicotinate
Methyl-6-methoxynicotinate
Methyl-6-methoxynicotinate
Irritant
Methyl-6-methoxynicotinate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl-6-methoxynicotinate
Supplier:
Apollo Scientific
About this item
CAS Number:
26218-80-4
:
Formula:
C₈H₉NO₃
MDL Number:
MFCD00674130
:
167.16 g/mol
Boiling Pt:
92 °C/3.5mmHg
Melting Pt:
48…52 °C
:
GHS07
Storage Temperature:
Ambient