Order Entry
Export
ContactUsLinkComponent
3-Chloro-o-xylene 98%
Irritant
3-Chloro-o-xylene 98%
Catalog # APOSOR15013-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Chloro-o-xylene 98%
Catalog # APOSOR15013-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :140.61 g/mol
Boiling Pt:194 °C
Density:1.1 g/cm³ (25 °C)
Flash Pt:73 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:608-23-1
Formula:C₈H₉Cl
MDL Number:MFCD00060689