Order Entry
Export
ContactUsLinkComponent
15-Hydroxypentadecanoic acid 99%
Irritant
15-Hydroxypentadecanoic acid 99%
Catalog # APOSOR1489-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
15-Hydroxypentadecanoic acid 99%
Catalog # APOSOR1489-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :258.40 g/mol
Melting Pt:84…89 °C
EINECS:225-026-1
 :GHS07
Storage Temperature:Ambient
CAS Number:4617-33-8
Formula:C₁₅H₃₀O₃
MDL Number:MFCD00039535