Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Thieno[2,3-d]pyrimidin-4-ylamine
Thieno[2,3-d]pyrimidin-4-ylamine
Thieno[2,3-d]pyrimidin-4-ylamine
Irritant
Thieno[2,3-d]pyrimidin-4-ylamine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Thieno[2,3-d]pyrimidin-4-ylamine
Supplier:
Apollo Scientific
About this item
CAS Number:
14080-56-9
:
Formula:
C₆H₅N₃S
MDL Number:
MFCD01316189
:
151.19 g/mol
Melting Pt:
234…236 °C
:
GHS07
Storage Temperature:
Ambient