Order Entry
Export
ContactUsLinkComponent
(S)-3-([1,1'-Biphenyl]-4-yl)-2-aminopropanoic acid 97%
(S)-3-([1,1'-Biphenyl]-4-yl)-2-aminopropanoic acid 97%
Catalog # APOSOR14744-250MG
Supplier:  Apollo Scientific
CAS Number:  
(S)-3-([1,1'-Biphenyl]-4-yl)-2-aminopropanoic acid 97%
Catalog # APOSOR14744-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :241.29 g/mol
Storage Temperature:Ambient
CAS Number:155760-02-4
Formula:C₁₅H₁₅NO₂
MDL Number:MFCD01318878