
2,4-Dichloro-D-phenylalanine
Catalog # APOSOR14718-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:234.09 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:114872-98-9
Formula:C₉H₉Cl₂NO₂
MDL Number:MFCD01860873