Order Entry
Export
ContactUsLinkComponent
Fmoc-2,4-dichloro-D-phenylalanine 97%
Fmoc-2,4-dichloro-D-phenylalanine 97%
Catalog # APOSOR14605-5G
Supplier:  Apollo Scientific
CAS Number:  
Fmoc-2,4-dichloro-D-phenylalanine 97%
Catalog # APOSOR14605-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :456.33 g/mol
Storage Temperature:Ambient
CAS Number:352351-61-2
Formula:C₂₄H₁₉Cl₂NO₄
MDL Number:MFCD01863047