
Ethyl imidazo[2,1-b]thiazole-6-carboxylate
Catalog # APOSOR1447-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:87…91 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:64951-04-8
Formula:C₈H₈N₂O₂S