Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-3,5-dibromo-2-methoxybenzenecarboxylate
Methyl-3,5-dibromo-2-methoxybenzenecarboxylate
Methyl-3,5-dibromo-2-methoxybenzenecarboxylate
APOSOR14437-1G
:
Apollo Scientific
:
Methyl-3,5-dibromo-2-methoxybenzenecarboxylate
APOSOR14437-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
323.98 g/mol
:
52…54 °C
:
GHS07
:
Ambient
:
15790-59-7
:
C₉H₈Br₂O₃
:
MFCD00189410