
Methyl-3-(1,3-oxazol-5-yl)benzoate, Technical Grade
Catalog # APOSOR1440-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:203.2 g/mol
Melting Pt:102…105 °C
:GHS07
Storage Temperature:Ambient
CAS Number:850375-14-3
Formula:C₁₁H₉NO₃
MDL Number:MFCD06797482