Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-3-(methoxycarbonyl)benzoic acid
2-Amino-3-(methoxycarbonyl)benzoic acid 97%
2-Amino-3-(methoxycarbonyl)benzoic acid 97%
APOSOR14348-250MG
:
Apollo Scientific
:
2-Amino-3-(methoxycarbonyl)benzoic acid 97%
APOSOR14348-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
195.1721 g/mol
:
170…172 °C
:
GHS07
:
Ambient
:
253120-47-7
:
C₉H₉NO₄
:
MFCD06200897