3-(Dimethylamino)phenol
Bottle, 1 g
Catalog # APOSOR14323-1G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: N,N-Dimethyl-3-hydroxyaniline, 3-Dimethylaminophenol
- CAS Number:99-07-0
- Formula:C₈H₁₁NO
- MDL Number:MFCD00002264
- Molecular Weight:137.18 g/mol
- Boiling Pt:267 °C
- Melting Pt:80…84 °C
- Flash Pt:148 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g



