
2,3-Dichloro-5,6-dicyano-p-benzoquinone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:84-58-2
:
Formula:C₈Cl₂N₂O₂
MDL Number:MFCD00001593
:227.01 g/mol
Melting Pt:210…215 °C
Storage Temperature:Ambient
EINECS:201-542-2
UN:3439
ADR:6.1,III
:GHS06