Order Entry
Export
ContactUsLinkComponent
5-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1H-indole
Irritant
5-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1H-indole
Catalog # APOSOR14268-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1H-indole
Catalog # APOSOR14268-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:179…181 °C
 :334.22 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:282546-99-0
Formula:C₁₆H₁₆BrNO₂
MDL Number:MFCD06411613