Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dimethyl-1,3-thiazol-2-amine
4,5-Dimethyl-1,3-thiazol-2-amine 95%
4,5-Dimethyl-1,3-thiazol-2-amine 95%
Irritant
4,5-Dimethyl-1,3-thiazol-2-amine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4,5-Dimethyl-1,3-thiazol-2-amine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
2289-75-0
:
Formula:
C₅H₈N₂S
MDL Number:
MFCD00462425
:
128.2 g/mol
Melting Pt:
66…69 °C
:
GHS07
Storage Temperature:
Refrigerator