
4-(4-Hydroxyphenyl)benzonitrile 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:19812-93-2
:
Formula:C₁₃H₉NO
MDL Number:MFCD00059625
:195.22 g/mol
Melting Pt:191…195 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07