
1-Phenyl-1,3,8-triazaspiro[4.5]decan-4-one 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1021-25-6
:
Formula:C₁₃H₁₇N₃O
MDL Number:MFCD00005977
:231.293 g/mol
Melting Pt:188…191 °C
EINECS:213-819-5
:GHS07
Storage Temperature:Refrigerator