Order Entry
Export
Orders LinkContactUsLinkComponent
3-Phenoxypropionic acid

3-Phenoxypropionic acid

Irritant
Supplier:   Apollo Scientific
3-Phenoxypropionic acid
Supplier:  Apollo Scientific
Certificates
3-Phenoxypropionic acid

3 Options Available Below

Product Details & Documents
Synonyms:  3-Penoxypropionic acid, 3-Phenoxypropanoic acid
  • CAS Number:7170-38-9
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00002769
  • Molecular Weight:166.18 g/mol
  • Boiling Pt:235…245 °C
  • Melting Pt:98…100 °C
  • Storage Temperature:Ambient
  • EINECS:230-518-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 57,90 €
    57,90 €
    1 * 1 g
       57,90 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 86,90 €
    86,90 €
    1 * 5 g
       86,90 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 232,00 €
    232,00 €
    1 * 25 g
       232,00 €