Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dimethoxy-2-nitrobenzoic acid
4,5-Dimethoxy-2-nitrobenzoic acid 99%
4,5-Dimethoxy-2-nitrobenzoic acid 99%
Irritant
4,5-Dimethoxy-2-nitrobenzoic acid 99%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4,5-Dimethoxy-2-nitrobenzoic acid 99%
Supplier:
Apollo Scientific
About this item
CAS Number:
4998-07-6
:
Formula:
C₉H₉NO₆
MDL Number:
MFCD00014697
:
227.17 g/mol
Melting Pt:
195…197 °C
:
GHS07
Storage Temperature:
Ambient