Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-2,6-diaminopyridine
4-Bromo-2,6-diaminopyridine
4-Bromo-2,6-diaminopyridine
:
Apollo Scientific
4-Bromo-2,6-diaminopyridine
:
Apollo Scientific
:
329974-09-6
:
:
C₅H₆BrN₃
:
MFCD04115308
:
188.03 g/mol
:
126 °C
:
GHS07
:
Ambient