Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dibromo-3-nitropyridine
2,6-Dibromo-3-nitropyridine
2,6-Dibromo-3-nitropyridine
APOSOR13599-5G
:
Apollo Scientific
:
2,6-Dibromo-3-nitropyridine
APOSOR13599-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
281.8900 g/mol
:
GHS07
:
Ambient
:
55304-80-8
:
C₅H₂Br₂N₂O₂
:
MFCD00233995