0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1-Methyl-2-oxo-1,2-dihydroquinoline-4-carbaldehyde
1-Methyl-2-oxo-1,2-dihydroquinoline-4-carbaldehyde
1-Methyl-2-oxo-1,2-dihydroquinoline-4-carbaldehyde
:
Apollo Scientific
1-Methyl-2-oxo-1,2-dihydroquinoline-4-carbaldehyde
:
Apollo Scientific
:
15112-98-8
:
:
C₁₁H₉NO₂
:
MFCD03942175
:
187.2 g/mol
:
GHS07
:
Ambient