Order Entry
Export
ContactUsLinkComponent
4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane
Irritant
4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane
Catalog # APOSOR13174-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane
Catalog # APOSOR13174-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :266.1410 g/mol
Melting Pt:60…62 °C
Storage Temperature:Freezer
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:680596-79-6
Formula:C₁₄H₂₃BO₄
MDL Number:MFCD04115656