Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Piperazine
Piperazine 97%
Piperazine 97%
:
Apollo Scientific
Piperazine 97%
:
Apollo Scientific
:
110-85-0
:
:
C₄H₁₀N₂
:
MFCD00005953
:
86.14 g/mol
:
145…148 °C
:
108…113 °C
:
1.1 g/cm³ (25 °C)
:
109 °C
:
Ambient
:
203-808-3
:
2579
:
8,III
:
GHS05
:
GHS07
:
GHS08