
1,3-Dibromobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:108-36-1
:
Formula:C₆H₄Br₂
MDL Number:MFCD00000078
:235.91 g/mol
Boiling Pt:218…220 °C
Melting Pt:–7 °C
Density:1.952 g/cm³ (25 °C)
Flash Pt:93 °C
EINECS:203-574-2
:GHS07
Storage Temperature:Ambient