1,4-Diethynylbenzene 97%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: p-Diethynylbenzene, 1,4-Diethylnylbenzene
- CAS Number:935-14-8
- Formula:C₁₀H₆
- MDL Number:MFCD00078375
- Molecular Weight:126.16 g/mol
- Melting Pt:92 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: p-Diethynylbenzene, 1,4-Diethylnylbenzene
- CAS Number:935-14-8
- Formula:C₁₀H₆
- MDL Number:MFCD00078375
- Molecular Weight:126.16 g/mol
- Melting Pt:92 °C
- Storage Temperature:Ambient



