Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(2-Thienyl)pyridine
2-(2-Thienyl)pyridine
2-(2-Thienyl)pyridine
Irritant
2-(2-Thienyl)pyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-(2-Thienyl)pyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
3319-99-1
:
Formula:
C₉H₇NS
MDL Number:
MFCD00044441
Melting Pt:
60…65 °C
:
161.22 g/mol
:
GHS07
Storage Temperature:
Refrigerator