

Isopentylamine
Catalog # APOSOR12816-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:87.16 g/mol
Boiling Pt:95…97 °C
Melting Pt:–60 °C
Density:0.751 g/cm³ (25 °C)
Flash Pt:–1 °C
Storage Temperature:Ambient
EINECS:203-528-0
UN:1160
ADR:3+8,II
:GHS05
:GHS07
CAS Number:107-85-7
Formula:C₅H₁₃N
MDL Number:MFCD00008203