Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(3,5-Dichlorophenyl)biguanide hydrochloride
1-(3,5-Dichlorophenyl)biguanide hydrochloride
1-(3,5-Dichlorophenyl)biguanide hydrochloride
Irritant
1-(3,5-Dichlorophenyl)biguanide hydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1-(3,5-Dichlorophenyl)biguanide hydrochloride
Supplier:
Apollo Scientific
About this item
CAS Number:
175205-04-6
:
Formula:
C₈H₁₀Cl₃N₅
Melting Pt:
251…255 °C
:
GHS07
Storage Temperature:
Ambient