0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
3',4'-Dichloro[1,1'-biphenyl]-4-carbaldehyde
3',4'-Dichloro[1,1'-biphenyl]-4-carbaldehyde
3',4'-Dichloro[1,1'-biphenyl]-4-carbaldehyde
APOSOR12654-1G
:
Apollo Scientific
:
3',4'-Dichloro[1,1'-biphenyl]-4-carbaldehyde
APOSOR12654-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
251.11 g/mol
:
95…98 °C
:
GHS07
:
Ambient
:
50670-78-5
:
C₁₃H₈Cl₂O
:
MFCD03002047