
2,5-Dibromopyrazine 97+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:23229-26-7
:
Formula:C₄H₂Br₂N₂
MDL Number:MFCD08061601
:237.8800 g/mol
Boiling Pt:234 °C
Density:2.197 g/cm³ (25 °C)
Flash Pt:95 °C
:GHS07
Storage Temperature:Ambient